ContactUsLinkComponent
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.53 mm, Length: 60 m
# de catalogue 76443-006
Fournisseur:  HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.53 mm, Length: 60 m
# de catalogue 76443-006
Fournisseur:  HICHROM LIMITED
Customers Who Bought This Also Bought