HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.32 mm, Length: 10 m
# de catalogue 76442-986
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.32 mm, Length: 10 m
# de catalogue 76442-986
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Fournisseur: VWR International
$50.87 to $493.91
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$72.06 to $624.50
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$73.75 to $568.62
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
$57.65 to $603.92
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$45.90 to $336.74
Target DP™ Glass Screw-Thread Vials, 9-42, Thermo Scientific
Fournisseur: NATIONAL SCIENTIFIC MX
$114.99 to $1,770.13
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$184.38
WHEATON® MicroLiter Crimp-Top Vials, 11 mm,Component Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$142.31
WHEATON® ABC Screw Caps, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$101.25 to $1,012.48
SureSTART™ Glass Crimp Top Vials, 2 ml, Level 2 High-Throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$1,347.98 to $1,347.98
WHEATON® MicroLiter 20 mm Crimp Seals with Septa, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$160.41
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
$1,853.47 to $1,853.47