HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.25 mm, Length: 5 m
# de catalogue 76442-970
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.25 mm, Length: 5 m
# de catalogue 76442-970
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Fournisseur: Advanced Instruments
$1,223.38
Cryo Markers, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$78.87 to $89.78
Sodium hydrogen carbonate ≥99%, powder USP, Multi-Compendial, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
$475.34 to $1,901.42
Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Fournisseur: Honeywell Research Chemicals
$1,359.73 to $2,133.93