HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.20 mm, Length: 20 m
# de catalogue 76442-962
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.20 mm, Length: 20 m
# de catalogue 76442-962
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® 11 mm Crimp Top Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$45.90 to $336.74
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$121.85 to $1,194.50
VWR® 11 mm Crimp Top Vials, Kits, and Inserts, Standard Opening
Fournisseur: VWR International
$40.01 to $399.86
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
$148.39
SureSTART™ Septa for 20 mm Stoppers, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$131.41 to $136.31
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$11,110.75 to $11,110.75
WHEATON® MicroLiter µLPlate® Component Kits, Inserts and Caps, Unassembled, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$184.38
WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$136.75
WHEATON® E-Z Vial® Large-Opening Crimp-Top Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$125.61