ContactUsLinkComponent
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.18 mm, Length: 20 m
# de catalogue 76442-950
Fournisseur:  HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.18 mm, Length: 20 m
# de catalogue 76442-950
Fournisseur:  HICHROM LIMITED
Customers Who Bought This Also Bought