HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.18 mm, Length: 20 m
# de catalogue 76442-950
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.18 mm, Length: 20 m
# de catalogue 76442-950
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
Exel International Brand Quality Catheter Tip Syringes, Air-Tite Products
Fournisseur: Air-Tite
63,91 $ to 263,95 $
Acetic acid glacial ≥99.7% for HPLC, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
358,83 $ to 1 481,22 $
VACUETTE® Sodium Citrate Coagulation Tubes, Greiner Bio-One
Fournisseur: Greiner Bio-One
18,45 $ to 442,53 $
MiniCollect* Tube K2E K2Edta Non sterile Lavender Cap 0.25/0.5 Ml PK500
Fournisseur: Greiner Bio-One
656,98 $ to 2 627,82 $
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Fournisseur: Electron Microscopy Science
90,42 $ to 94,61 $
Dow Corning® Vacuum Grease, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
113,37 $