HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.15 mm, Length: 10 m
# de catalogue 76442-936
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.15 mm, Length: 10 m
# de catalogue 76442-936
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® 10 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$127.69
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$135.22
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
Fournisseur: VWR International
$81.56 to $628.06
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$178.96
Screw Caps with Partial Slit Septa for 12×32 mm Vials, Microsolv Technology Corp
Fournisseur: MicroSolv Technology Corp
$119.01
WHEATON® MicroLiter Septa for 20 mm Crimp Seals, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$205.86
SureSTART™ Glass Screw Top 0.3 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$182.38 to $196.03
SureSTART™ Manual Crimper for 20 mm Crimp Vials, Level 3, Thermo Scientific
Fournisseur: Thermo Scientific
$634.78
SureSTART™ Crimp Caps, 20 mm, Level 2 High-throughput Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$180.01 to $182.41