HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.05 mm, Length: 5 m
# de catalogue 76442-916
Fournisseur: HICHROM LIMITED
HMDS retention gap/precolumn, high temperature, fused silica, Deactivation mainly for apolar solvents, Internal Diameter: 0.05 mm, Length: 5 m
# de catalogue 76442-916
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$132.38
Screw Top Vials and Caps, Easy Purchase Packs 9 mm, MicroSolv
Fournisseur: MicroSolv Technology Corp
$163.94
WHEATON® ABC Vial™ Large-Opening Screw-Thread Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$175.93
SureSTART™ Polypropylene Screw Top 2 ml Microvials for <2 ml Samples, Level 1 Everyday Analysis, Thermo Scientific
Fournisseur: Thermo Scientific
$1,853.47 to $1,853.47
SureSTART™ Total Recovery Glass Screw Top 1.5 ml Microvials for <2 ml Samples, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$4,158.43 to $4,158.43