DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.53 mm, Length: 60 m
# de catalogue 76442-910
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought