ContactUsLinkComponent
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.20 mm, Length: 5 m
# de catalogue 76442-862
Fournisseur:  HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.20 mm, Length: 5 m
# de catalogue 76442-862
Fournisseur:  HICHROM LIMITED
Customers Who Bought This Also Bought