ContactUsLinkComponent
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.18 mm, Length: 30 m
# de catalogue 76442-856
Fournisseur:  HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.18 mm, Length: 30 m
# de catalogue 76442-856
Fournisseur:  HICHROM LIMITED
Customers Who Bought This Also Bought