DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.15 mm, Length: 30 m
# de catalogue 76442-844
Fournisseur: HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.15 mm, Length: 30 m
# de catalogue 76442-844
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
WHEATON® MicroLiter Crimp-Top Headspace Vials, 20 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
109,95 $
SureSTART™ Glass Screw Top Vials, 2 ml, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
43,20 $ to 44,79 $
WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
166,99 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $