Toolbox K-Kit Excludes K-Kits Silver, K-kits are sample holders designed to for convenient TEM observation of liquid samples, allowing nano-objects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized, With vacuum compatible sealing of liquid in electron-transmitting thickness
# de catalogue 76439-818
Fournisseur: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.