ContactUsLinkComponent
Probe Kit, Material: Nylon Plastic, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 deg C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples
# de catalogue 76439-474
Probe Kit, Material: Nylon Plastic, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 deg C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples
Probe Kit, Material: Nylon Plastic, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 deg C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples
# de catalogue 76439-474
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.