Magic wand Curved/flat strong tip, Material: nylon, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 deg C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples.
# de catalogue 76439-470
Fournisseur: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.