Carbon PEEK Probes Kit, Material: Plastic, Made from PEEK high performance plastic with chemical resistantance, Withstands temperatures to 300 deg C, For electronic areas to labss, from preparation of samples to applying adhesives, Use as a stirring rod or dosing samples
# de catalogue 76439-460
Fournisseur: ELECTRON MICROSCOPY SCIENCE
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.