DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.10 mm, Length: 2 m
# de catalogue 76442-826
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought