DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 5 m
# de catalogue 76442-820
Fournisseur: HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 5 m
# de catalogue 76442-820
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
45,27 $
VWR® Screw Caps for 8-425 Screw-Thread Vials, Standard Opening
Fournisseur: VWR International
120,75 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
57,65 $ to 603,92 $
Screw Top Vials and Caps, Easy Purchase Packs 9 mm, MicroSolv
Fournisseur: MicroSolv Technology Corp
129,58 $
WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
723,90 $
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
Fournisseur: MicroSolv Technology Corp
438,81 $
Virgin PTFE liner only, 0.01 inch (10 mil thick), Diameter: 9.5mm
Fournisseur: MicroSolv Technology Corp
137,30 $