DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 2 m
# de catalogue 76442-818
Fournisseur: HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 2 m
# de catalogue 76442-818
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
VWR® Screw-Thread Vials 8-425, Caps and Inserts with Standard Opening
Fournisseur: VWR International
$64.11 to $458.29
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Fournisseur: VWR International
$118.61 to $1,128.11
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications, Thermo Scientific
Fournisseur: Thermo Scientific
$120.60 to $125.07
WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$452.40