DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 1 m
# de catalogue 76442-816
Fournisseur: HICHROM LIMITED
DPTMDS retention gap/precolumn, high temperature, fused silica, General purpose deactivation for both apolar and polar solvents, Internal Diameter: 0.05 mm, Length: 1 m
# de catalogue 76442-816
Fournisseur: HICHROM LIMITED
Customers Who Bought This Also Bought
WHEATON® MicroLiter Screw Caps for Vials, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
91,98 $
SureSTART™ Screw Caps, 9 mm, Level 3 High Performance Applications
Fournisseur: Thermo Scientific
79,21 $ to 82,14 $
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Fournisseur: VWR International
86,08 $ to 717,60 $
Screw Top Vials and Caps, Easy Purchase Packs 9 mm, MicroSolv
Fournisseur: MicroSolv Technology Corp
129,58 $
WHEATON® MicroLiter Sample Vial Assembled Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
166,99 $
MicroSolv™ Pharma Grade Vials, Caps and Convenience Kits
Fournisseur: MicroSolv Technology Corp
52,50 $
WHEATON® MicroLiter Snap-Top Vials, 11 mm, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
231,64 $
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
118,60 $