ContactUsLinkComponent
VWR* High and consistent recoveries, Fast and easy sample preparation and method development, vwr* Cartridge, Retain PEP/CX/AX SPE, Versatile polymeric materials for retention of polar, non-polar, basic and acidic drugs, Exceptional recoveries for polar and non-polar analytes, Volume: 6ml
# de catalogue 76439-334
Fournisseur:  VWR International
VWR* High and consistent recoveries, Fast and easy sample preparation and method development, vwr* Cartridge, Retain PEP/CX/AX SPE, Versatile polymeric materials for retention of polar, non-polar, basic and acidic drugs, Exceptional recoveries for polar and non-polar analytes, Volume: 6ml
# de catalogue 76439-334
Fournisseur:  VWR International
Customers Who Bought This Also Bought