Mask, 3 layers of polypropylene with two elastic silicon-free earloop bands. Material is soft to help prevent irritation. Includes nose piece to keep secure. Use in pharmaceutical, biotech, LAR, and healthcare settings.
# de catalogue 76437-404
Fournisseur: Veltek Associates
Customers Who Bought This Also Bought