Empty Racks for Pipette Tips, Socorex® Qualitix® Premium, DWK Life Sciences
# de catalogue 76437-362
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Empty tip racks
- For Use With:1000 µl tips
Spécifications
A propos de cet article
Empty racks for tip refilling.
- Superior quality, autoclavable
- Lot by lot consistency
- Certified free of nuclease, endotoxin and ATP
- Reduced packaging size, less bench space requirement
- Broad compatibility with major micropipette brands
Customers Who Bought This Also Bought