PLR SPE 96-well plate, 25 mg/2 mL each well, for high-throughput or automated workflows, Remove both proteins and phospholipids from biological samples in one easy, high-efficiency procedure, Avoid signal suppression by removing interfering phospholipids from the sample matrix, No method development
# de catalogue 76437-260
Fournisseur: Restek
Customers Who Bought This Also Bought