Vial, Limited-Volume, 9 mm Screw-Thread, Polypropylene, Fit all 2.0 mL, 12 x 32 mm, vial-based autosamplers, PTFE-free, ideal for PFAS analysis and other PFAS-sensitive methods, Color: Clear, Size:12 x 32 mm, Volume:700uL
# de catalogue 76437-250
Fournisseur: Restek
Customers Who Bought This Also Bought