Acrylamide-d3, 500 ug/mL in acetonitrile, 5 mL/ampul, Deuterium-labeled acrylamide is an excellent and cost-effective isotopically labeled internal standard choice for the analysis of acrylamide in food or environmental samples, Storage Temperature:10 deg C or colder
# de catalogue 76437-236
Fournisseur: Restek
Customers Who Bought This Also Bought