ContactUsLinkComponent
BOD Overcap, use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods, Our overcaps are made of polyethylene and fit snugly against the rim and shoulder of our disposable BOD bottles, These overcaps do NOT fit the glass BOD bottles, for Bottle
Accessories for B.O.D Disposable Bottles, Environmental Express
Catalog # 76436-870
undefined
Accessories for B.O.D Disposable Bottles, Environmental Express
Catalog # 76436-870
Specifications
  • Description:
    Overcaps for B.O.D bottles, PE
  • Cat. no.:
    76436-870
Specifications
About this item
These overcaps are made of PE and fit snugly against the rim and shoulder of the disposable BOD bottles.
  • Ideal for 300 ml BOD bottles
The use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods, and these overcaps do not fit the glass BOD bottles.
Frequently Bought Together