Accessories for B.O.D Disposable Bottles, Environmental Express
Catalog # 76436-870
Supplier: Environmental Express
Specifications
- Description:Overcaps for B.O.D bottles, PE
- Cat. no.:76436-870
Specifications
About this item
These overcaps are made of PE and fit snugly against the rim and shoulder of the disposable BOD bottles.
- Ideal for 300 ml BOD bottles
The use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods, and these overcaps do not fit the glass BOD bottles.
Frequently Bought Together