Gloss Meter, TG 268, High End, Flexible, Handheld Gloss Meter - 20 / 60 / 85 Measuring Angle, Colour Type: Opaque, Display: TFT 3.5 inch, capacitive touch screen, Interfaces: USB, RS 232, Portable, Power Supply: 3200 mAh Li-ion Battery, Operating Temperature 0-40 C, Dimensions: 160 x 90 x 75 mm
# de catalogue 76424-354
Fournisseur: Lovibond
Customers Who Bought This Also Bought