Drip Trough, under mount style, Material: stainless steel construction, Made from 20 gauge material, no. 4 finish, 3/4inch mounting flange, 1inch deep trough, Dimension: 30X4in
# de catalogue 76439-310
Fournisseur: Interdyne
Drip Trough, under mount style, Material: stainless steel construction, Made from 20 gauge material, no. 4 finish, 3/4inch mounting flange, 1inch deep trough, Dimension: 30X4in
# de catalogue 76439-310
Fournisseur: Interdyne
Customers Who Bought This Also Bought
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with 10-Button Direct Input and Certificate of Calibration, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$48.82 to $442.63
VWR® Single Channel Electronic Timer with Memory and Clock and Certificate of Calibration
Fournisseur: VWR International
$75.03 to $739.18
VWR® Single Channel Electronic Timer with Memory and Certificate of Calibration
Fournisseur: VWR International
$77.73 to $1,082.63
SP Bel-Art H-B® DURAC® Single-Channel Electronic Timer with Certificate of Calibration, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
$31.17 to $282.75
Shimpo Non-Contact Pocket Laser Tachometers, Nidec Shimpo America
Fournisseur: Nidec Shimpo America
$368.65