Pegboard, V Series, have a catch drain and integral drip trough in 2in or 4in depth, All drip troughs some with a 36in clear flexible drain tube, white Polypropylene pegs, Autoclavable, Constructed from 20 gauge, type 304 stainless steel, polished to a no. 4 finish, Wall mount bracket shipped
# de catalogue 76439-286
Fournisseur: Interdyne
Customers Who Bought This Also Bought