Microtube Holder, Horizontal Plastic Clip, holds up to 24 X 1.5 / 2.0mL microtubes, For use on any Vortex-Genie 2 or Vortex-Genie Pulse brand vortex mixer when doing bead beating, extractions, or just need more aggressive hands-free vortexing of microtubes
# de catalogue 76424-338
Fournisseur: Scientific Industries
Customers Who Bought This Also Bought