Spécifications
- Membrane Material:Polyethersulfone (PES)
- Max. RCF:10000 ×g
- Ø×H:17×122 mm
- Dead volume:<10 µl
- Filtration area:2.5 cm²
- Packaging:25
- MWCO:10 kD
Spécifications
A propos de cet article
Vivaspin® 6 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin® 6 can process 6 ml in either swing bucket or fixed angle rotors accepting standard 15 ml conical bottom test tubes.
- High retentate recovery <90%
- Vertical membrane design for ultra-fast concentration
- Safe sample concentration due to dead-stop volume
- Convenient concentrate recovery by pipetting
- Volume graduation for easy reading/monitoring
Vivaspin® 6 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. The remaining volume is easy to read off the printed scale on the side of the concentrator and the modified dead stop pocket further simplifies direct pipette recovery of the final concentrate.
Vivaspin® 6 is optimally suited for the concentration of antibody-, protein-, nanoparticle- and virus samples from 6 ml sample volume to 30 µl in a centrifuge accepting 50 ml conical bottom centrifuge tubes. These are the only device of its size that can operate 6 ml sample.