Order Entry
ContactUsLinkComponent
Strontium
Assurance® Grade Strontium Standards for AA and ICP, SPEX CertiPrep
# de catalogue SXPLSR2-2M
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Assurance® Grade Strontium Standards for AA and ICP, SPEX CertiPrep
# de catalogue SXPLSR2-2M
Fournisseur:  SPEX CERTIPREP LLC
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Concentration:
    1000 µg/ml
  • Element:
    Strontium
  • Matrix:
    2% HNO₃
  • Packaging:
    LDPE bottle
  • Size:
    30 ml

Spécifications

A propos de cet article

Single-element standards for AA & ICP.

  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
  • Directly traceable to NIST (where applicable)

AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.

Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Customers Who Bought This Also Bought