Assurance® Grade Arsenic Standards for AA and ICP, SPEX CertiPrep
Catalogue # SXPLAS2-2Y
Fournisseur: SPEX CERTIPREP LLC
Numéro CAS:
Spécifications
- Concentration:1000 µg/ml
- Matrix:2% HNO₃
- Pack type:LDPE bottle
- Size:125 ml
- Cat. no.:SXPLAS2-2Y
- Element:Arsenic
Spécifications
À propos de cet article
Single-element standards for AA & ICP.
- Made with acid and ASTM Type I Water
- Inorganic compounds and metals at 99.99% to 99.9999% purity (where commercially available)
- Directly traceable to NIST (where applicable)
AA & ICP Assurance® Grade CRMs are designed for AA and ICP and are available in single and multi-element formulations. 70 elements are available as single-element standards and are available at 1,000 µg/ml and/or 10,000 µg/ml. They are available packaged in 30 ml, 125 ml, 250 ml, and 500 ml bottles to minimize contamination.
Certifications: Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Frequently Bought Together