Order Entry
Canada
ContactUsLinkComponent
PolarSafe® Cooling Transport Bags, Cole Parmer
Catalogue # 76420-932
Fournisseur:  Antyila Scientific
undefined
PolarSafe® Cooling Transport Bags, Cole Parmer
Catalogue # 76420-932
Fournisseur:  Antyila Scientific
Supplier Number:  03726-03

Spécifications

  • Capacity:
    30 L
  • Color:
    Black/Blue
  • Description:
    Cole-Parmer PolarSafe® transport bag
  • Dimensions:
    410×240×350 mm
  • Cat. no.:
    76420-932

Spécifications

À propos de cet article

These insulated nylon bags provide an easy solution for transporting your samples.

  • Maintain specific temperature ranges for extended periods of time for improved sample protection
  • Get the temperature protection you need with choice of durable transport bags or boxes
  • Modular, color-coded cooling packs allow flexible configurations for different-sized samples
  • Ensure sample integrity with optional security sealing ties

Organize cooling packs to suit your sample or choose from one of our prepared kits. Bags have pockets to keep documentation safe during transit, a shoulder strap, and carrying handles.

Frequently Bought Together