Specifications
- Volume:Microbore, low flow
- Description:Sterile dual Luered extension lines
- Length:36"
- Material:PVC
- Cat. no.:76420-260
Specifications
About this item
SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing, extension lines include one male and one female luer adaptor, and are available in a variety of standard lengths.
- Lines feature one male luer end and one female luer end
- ETO sterilized
- High flow and micro flow options available
- Custom sizes are also available
SAI's luered extension lines are compatible with any standard luer fitting, providing a simple, quick, and secure luer connection. These luered fittings offer unlimited flexibility for research and infusion needs. Designed for animal research use only.