Order Entry
ContactUsLinkComponent
Bottle Top Dispenser, Volume Range: 0.25 - 2.5 mL, Tread GL32
Corning® Bottle Top Dispensers, Corning
# de catalogue 76420-190
Fournisseur:  Corning
Bottle Top Dispenser, Volume Range: 0.25 - 2.5 mL, Tread GL32
Corning® Bottle Top Dispensers, Corning
# de catalogue 76420-190
Fournisseur:  Corning

Spécifications

  • Volume:
    0.25 - 2.5 ml
  • Cat. no.:
    76420-190
  • Neck Size:
    32 mm
  • Increments:
    0.05 ml

Spécifications

A propos de cet article

Corning® bottle top dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent. Depending on the model, they can accommodate volumes from 0.25 to 100 ml.

  • Accurate and reproducible volume setting
  • Fully autoclavable for protection against contamination
  • Rotating valve block for convenient dispensing
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system for user safety at work
  • Easy in-lab calibration with built-in recalibration mechanism

In-lab calibration tool and set of four thread adapters included with every product.

Certifications: Meets ISO 8655 requirements.
Delivery: Dispensers up to 10 ml are delivered with adapter sizes: A25, A28, A40, A45. Dispensers up to 25, 50, and 100 ml are delivered with adapter sizes: A25, A28, A38, A45/32.
Customers Who Bought This Also Bought