VWR® Flat Caps for Screw Cap Microtubes
# de catalogue 76417-218
Fournisseur: VWR International
Spécifications
- Sterility:Non sterile
- Cat. no.:76417-218
- Cap Color:Opaque blue
Spécifications
A propos de cet article
Flat caps with O-ring for use with VWR® screw cap microtubes only (series 76417-xxx).
Feature ³/₄ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Customers Who Bought This Also Bought