ContactUsLinkComponent
Microplate
SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
# de catalogue 76415-938
Fournisseur:  Thermo Scientific
SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
# de catalogue 76415-938
Fournisseur:  Thermo Scientific
Spécifications
  • Volume:
    220 µl
  • Description:
    SureSTART™ WebSeal™ 96-well microtiter plates, round U, V, and flat-bottom, Level 3 high-throughput applications
  • Material:
    Polypropylene
  • I.D.×H:
    7×14.7 mm
  • Well Shape:
    Round, conical bottom (V-well)
Spécifications
A propos de cet article
SureSTART™ Level 3 WebSeal™ Plate 96-well round U, V, and flat-bottom microtiter plates are designed for regulated labs that require high performing products. Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
  • Specification certified
  • Suitable for HPLC/UHPLC applications
  • Performance Level 3 products include unique WebSeal Plate+ glass-coated polypropylene plates and plates with glass inserts
  • Suitable for QA/QC applications using core detectors such as UV, DAD/PDA, RI, fluorescence, ELSD, and MS analysis
Plate+ glass-coated Level 3 well plates are recommended for use with very sensitive applications, low sample concentrations, and hydrophobic samples such as proteins and are ideal for use with all solvent types including non-polar solvents.
Customers Who Bought This Also Bought