SureSTART™ WebSeal™ Well Microtiter Plates, Round U, V and Flat-Bottom
# de catalogue 76415-938
Fournisseur: Thermo Scientific
Spécifications
- Volume:220 µl
- Description:SureSTART™ WebSeal™ 96-well microtiter plates, round U, V, and flat-bottom, Level 3 high-throughput applications
- Material:Polypropylene
- I.D.×H:7×14.7 mm
- Well Shape:Round, conical bottom (V-well)
Spécifications
A propos de cet article
SureSTART™ Level 3 WebSeal™ Plate 96-well round U, V, and flat-bottom microtiter plates are designed for regulated labs that require high performing products. Level 3 well plates are for high performance applications and are suitable for regulated labs using all solvent types including non-polar solvents.
- Specification certified
- Suitable for HPLC/UHPLC applications
- Performance Level 3 products include unique WebSeal Plate+ glass-coated polypropylene plates and plates with glass inserts
- Suitable for QA/QC applications using core detectors such as UV, DAD/PDA, RI, fluorescence, ELSD, and MS analysis
Plate+ glass-coated Level 3 well plates are recommended for use with very sensitive applications, low sample concentrations, and hydrophobic samples such as proteins and are ideal for use with all solvent types including non-polar solvents.
Customers Who Bought This Also Bought