ContactUsLinkComponent
Test Strips, Nitrate/Nitrite, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85439.601
Fournisseur:  VWR International
Numéro CAS:  
undefined
Test Strips, Nitrate/Nitrite, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85439.601
Fournisseur:  VWR International
Numéro CAS:  
Spécifications
  • Description:
    Nitrate test strips 0 - 500mg/l
    Nitrite test strips 0 - 80 mg/l
  • Cat. no.:
    BDH85439.601
  • Range:
    0 · 10 · 25 · 50 · 100 · 250 · 500
    0 · 1 · 5 · 10 · 20 · 40 · 80
  • No. of Tests:
    100
Spécifications
À propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6 x 95 mm.
Frequently Bought Together