Test Strips, Nitrate/Nitrite, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85439.601
Fournisseur: VWR International
Numéro CAS:
Spécifications
- Description:Nitrate test strips 0 - 500mg/l
Nitrite test strips 0 - 80 mg/l - Cat. no.:BDH85439.601
- Range:0 · 10 · 25 · 50 · 100 · 250 · 500
0 · 1 · 5 · 10 · 20 · 40 · 80 - No. of Tests:100
Spécifications
À propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6 x 95 mm.
Frequently Bought Together