Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85434.601
Fournisseur: VWR International
Numéro CAS:
Spécifications
- Description:Peroxide test strips 0 - 100 mg/l
- Cat. no.:BDH85434.601
- Range:0 · 1 · 3 · 10 · 30 · 100
- No. of Strips:100
Spécifications
À propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
- Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.
Frequently Bought Together