ContactUsLinkComponent
Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85434.601
Fournisseur:  VWR International
Numéro CAS:  
undefined
Test Strips, Peroxide, Dosatest, VWR Chemicals BDH®
Catalogue # BDH85434.601
Fournisseur:  VWR International
Numéro CAS:  
Spécifications
  • Description:
    Peroxide test strips 0 - 100 mg/l
  • Cat. no.:
    BDH85434.601
  • Range:
    0 · 1 · 3 · 10 · 30 · 100
  • No. of Strips:
    100
Spécifications
À propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
Packaging: Container with 100 test strips 6×95 mm.
Frequently Bought Together