Spécifications
- Description:Peroxide test strips 0 - 25 mg/l
- Cat. no.:BDH85433.601
- Range:0 · 0,5 · 2 · 5 · 10 · 25
- No. of Strips:100
Spécifications
A propos de cet article
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
- Choice of ranges for required precision
Ideal to monitor your desinfection processes. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.