Gloves, disposable, Colour: Green, best chemical protection in disposable range, the compromise between protection and dexterity, has a high tear resistence and will not break when removing from box or donning the glove, Size: 9.5in length, 4mil, size 7
# de catalogue 76415-006
Fournisseur: MAPA
Customers Who Bought This Also Bought