RC 5 control, Temperature Control, Powerful recirculating chiller, designed for external cooling applications up to -30 deg C, Speed-controlled refrigeration system with 1400 W cooling capacity (20C), Natural thermofluid R290, Extended operating temperature range until 80 C, Digital level indicator
# de catalogue 76414-130
Fournisseur: IKA Works
Customers Who Bought This Also Bought