ContactUsLinkComponent
Filter, Sterile, accessory for Macroman pipette controller, The filter type used is a hydrophobic membrane filter (Female Luer-Lock inlet, Male Luer-Slip outlet), Size: 0.22 um
Accessories for MACROMAN™ Pipette Controller, Gilson
# de catalogue 76414-016
Fournisseur:  GILSON, INC.
Accessories for MACROMAN™ Pipette Controller, Gilson
# de catalogue 76414-016
Fournisseur:  GILSON, INC.
Spécifications
  • Description:
    Filter, 0.22 micrometer, sterile (set of 10) for MACROMAN
  • Cat. no.:
    76414-016