Shelf, reinforced - positioned at bottom level, max. Load 67 kg, with additional attachment for operation of shaking device, stirring device or roller bottle system, for Growth chambers with light and humidity, Model KBWF 720
# de catalogue 76410-150
Fournisseur: Binder
Customers Who Bought This Also Bought