Alarm output, zero-voltage - for temperature (+/-2 degree C) and humidity (+/-5 % RH), accessible via 6-pin DIN socket (max. 24 V - 2.5 A), with audible signal that can be switched off, for model KBWf 240(Growth chamber)
# de catalogue 76410-128
Fournisseur: Binder
Customers Who Bought This Also Bought