Analog output 4-20 mA - for temperature and humidity values (output not adjustable), for Model KBWf 240(Growth chamber)
# de catalogue 76410-120
Fournisseur: Binder
Analog output 4-20 mA - for temperature and humidity values (output not adjustable), for Model KBWf 240(Growth chamber)
# de catalogue 76410-120
Fournisseur: Binder
Customers Who Bought This Also Bought
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Fournisseur: VWR International
$69.94
Shimpo LED Contact Handheld Tachometers, Nidec Shimpo America
Fournisseur: Nidec Shimpo America
$649.11
Shimpo Compact LCD Contact and Contact/Non-Contact Tachometers, Nidec Shimpo America
Fournisseur: Nidec Shimpo America
$518.81