ContactUsLinkComponent
RS 485 interface, 2-wire - Additional serial interface can be used parallel to Ethernet, for Multi Management Software APT-COM, for model KBW240(Growth chambers with light and humidity)
# de catalogue 76410-048
Fournisseur:  Binder
RS 485 interface, 2-wire - Additional serial interface can be used parallel to Ethernet, for Multi Management Software APT-COM, for model KBW240(Growth chambers with light and humidity)
# de catalogue 76410-048
Fournisseur:  Binder
Customers Who Bought This Also Bought