ContactUsLinkComponent
Dissecting Pan Aluminum with Wax, Dimensions (L x W x H): 13X9.5X2In, Color: Silver tray, Black wax
Aluminum Dissecting Trays with Wax
# de catalogue 470325-656
Dissecting Pan Aluminum with Wax, Dimensions (L x W x H): 13X9.5X2In, Color: Silver tray, Black wax
Aluminum Dissecting Trays with Wax
# de catalogue 470325-656
Spécifications
  • Description:
    Dissecting pan, large with black wax
  • Cat. no.:
    470325-656
  • L×W×H:
    33×24×5 cm
Spécifications
A propos de cet article
Made of aluminum and filled with a layer of paraffin wax, these dissecting pans will give you years of service with minimal maintenance.
  • Easy to Clean
  • Ideal Size for Smaller Specimens
  • Easy to Store
The paraffin holds T-pins securely without breaking down with repeated use. The wax resists absorption of most solutions allowing for a simple rinse and pour clean-up. This size of tray is ideal for smaller dissections like Clams, Squid, earthworm, locust, and most perch or smaller frogs.
Refill wax is available.
Customers Who Bought This Also Bought