Aluminum Dissecting Trays with Wax
# de catalogue 470325-656
Fournisseur: United Scientific Supplies
Spécifications
- Description:Dissecting pan, large with black wax
- Cat. no.:470325-656
- L×W×H:33×24×5 cm
Spécifications
A propos de cet article
Made of aluminum and filled with a layer of paraffin wax, these dissecting pans will give you years of service with minimal maintenance.
- Easy to Clean
- Ideal Size for Smaller Specimens
- Easy to Store
The paraffin holds T-pins securely without breaking down with repeated use. The wax resists absorption of most solutions allowing for a simple rinse and pour clean-up. This size of tray is ideal for smaller dissections like Clams, Squid, earthworm, locust, and most perch or smaller frogs.
Refill wax is available.
Customers Who Bought This Also Bought