Bottles with Dropper, Boston Round, Amber, United Scientific Supplies
# de catalogue 470325-636
Fournisseur: United Scientific Supplies
Spécifications
- Capacity:30 ml
- Color:Amber
- Description:Bottles with dropper, Boston round
- Cat. no.:470325-636
Spécifications
A propos de cet article
Amber Boston round bottle with droppers for general use.
- Screw cap with dropper assembly included
- Non-porous glass is easy to clean and autoclavable
- Amber color protects light-sensitive contents
These amber Boston round flint glass bottles include screw cap with dropper assembly are designed for general use. The glass is non-porous, which allows for easy cleaning or autoclaving. The amber color helps protect light sensitive materials.
Customers Who Bought This Also Bought